TCIAD2736

D2736-ZONE
TCD2736-ZONEEA 962.83 USD
TCD2736-ZONE
TCIAD2736
CAS Number: 1625-91-8
MDL Number: MFCD00008834
Molecular Formula: C20H26
Molecular Weight: 266.43
Boiling point (°C): 192
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR