2,6-Dimethoxy-p-benzoquinone ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,6-Dimethoxy-1,4-Benzoquinone

D2706-5G D2706-25G
TCD2706-025GEA 157.62 USD
TCD2706-025G TCD2706-005G
2,6-Dimethoxy-p-benzoquinone ≥97.0% (by HPLC)
2,6-Dimethoxy-p-benzoquinone

CAS Number: 530-55-2
MDL Number: MFCD00016368
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 2,6-Dimethoxy-p-quinone

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00016368
CAS Number: 530-55-2
EINECS: 208-484-7
https://us.vwr.com/stibo/web/std.lang.all/98/13/28979813.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR