2H,3H-Perfluoropentane ≥98.0%
|
Warning
|
Synonyms:
2H,3H-Decafluoropentane
CAS Number: 138495-42-8
MDL Number: MFCD00798129
Molecular Formula: C5H2F10
Molecular Weight: 252.06
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.60
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry
Other Product Names / Synonyms: 2H,3H-Perfluoropentane, 1,1,1,2,3,4,4,5,5,5-Decafluoropentane
Formula:
C₅H₂F₁₀ MW: 252.05 g/mol Density: 1.60 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00798129 CAS Number: 138495-42-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...