3,5-Dichlorophenylhydrazinium chloride ≥90.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3,5-Dichlorophenylhydrazine hydrochloride
, 1-(3,5-Dichlorophenyl)hydrazine hydrochloride
D2644-25G
D2644-5G
TCD2644-005GEA
31.42
USD
TCD2644-005G
TCD2644-025G
3,5-Dichlorophenylhydrazinium chloride ≥90.0% (by HPLC, titration analysis)
3,5-Dichlorophenylhydrazinium chloride
CAS Number: 63352-99-8
MDL Number: MFCD00012938
Molecular Formula: C6H6Cl2N2
Molecular Weight: 213.49
Purity/Analysis Method:
Form: Crystal
Formula:
C₆H₇Cl₃N₂ MW: 213.49 g/mol |
MDL Number:
MFCD00012938 CAS Number: 63352-99-8 EINECS: 264-109-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...