Dibenzyl diisopropylphosphoramidite ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Dibenzyl N,N-diisopropylphosphoramidite
CAS Number: 108549-23-1
MDL Number: MFCD00191988
Molecular Formula: C20H28NO2P
Molecular Weight: 345.42
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 70
Specific Gravity (20/20): 1.04
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Dibenzyloxy(diisopropylamino)phosphine
Formula:
C₂₀H₂₈NO₂P MW: 345.43 g/mol Density: 1.04 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD00191988 CAS Number: 108549-23-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...