2,5-Diiodothiophene ≥97.0%

Supplier: TCI America

D2611-5G
TCD2611-005GEA 95 USD
TCD2611-005G
2,5-Diiodothiophene ≥97.0%
2,5-Diiodothiophene

CAS Number: 625-88-7
MDL Number: MFCD00014525
Molecular Formula: C4H2I2S
Molecular Weight: 335.93
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 41
Flash Point (°C): 96
Storage Temperature: 0-10°C

Formula: C₄H₂I₂S
MW: 335.93 g/mol
Boiling Pt: 140 °C
Melting Pt: 41 °C
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014525
CAS Number: 625-88-7
EINECS: 210-915-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR