2,6-Dichloro-5-fluoronicotinic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

D2610-5G D2610-25G
TCD2610-005GEA 20.3 USD
TCD2610-005G TCD2610-25G
2,6-Dichloro-5-fluoronicotinic acid ≥97.0% (by titrimetric analysis)
2,6-Dichloro-5-fluoronicotinic acid

CAS Number: 82671-06-5
MDL Number: MFCD00799517
Molecular Formula: C6H2Cl2FNO2
Molecular Weight: 209.99
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 153


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Dichloro-5-fluoro-3-pyridinecarboxylic Acid

Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00799517
CAS Number: 82671-06-5
https://us.vwr.com/stibo/web/std.lang.all/95/80/28979580.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR