2,5-Dibromotoluene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,4-Dibromo-2-methylbenzene
D2577-5G
TCD2577-005GEA
25.38
USD
TCD2577-005G
2,5-Dibromotoluene ≥98.0%
2,5-Dibromotoluene
CAS Number: 615-59-8
MDL Number: MFCD00000090
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 113
Freezing point (°C): 7
Specific Gravity (20/20): 1.85
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 236 °C (1013 hPa) Melting Pt: 6 °C Density: 1.85 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00000090 CAS Number: 615-59-8 EINECS: 210-437-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...