2,6-Dibromo-4-fluoroaniline ≥98.0%
Supplier: TCI America
D2570-25G
TCD2570-025GEA
325
USD
TCD2570-025G
2,6-Dibromo-4-fluoroaniline ≥98.0%
2,6-Dibromo-4-fluoroaniline
CAS Number: 344-18-3
MDL Number: MFCD00041445
Molecular Formula: C6H4Br2FN
Molecular Weight: 268.91
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 66
Formula:
C₆H₄Br₂FN MW: 268.91 g/mol Boiling Pt: 277 °C (1013 hPa) Melting Pt: 66 °C |
MDL Number:
MFCD00041445 CAS Number: 344-18-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...