1,3-Difluoro-4-iodobenzene ≥98.0%
|
Warning
|
Synonyms:
2,4-Difluoro-1-iodobenzene
, 2,4-Difluoroiodobenzene
D2545-25G
TCD2545-025GEA
93.73
USD
TCD2545-025G
1,3-Difluoro-4-iodobenzene ≥98.0%
1,3-Difluoro-4-iodobenzene
CAS Number: 2265-93-2
MDL Number: MFCD00040151
Molecular Formula: C6H3F2I
Molecular Weight: 239.99
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 176
Flash Point (°C): 68
Specific Gravity (20/20): 2.00
Formula:
C₆H₃F₂I MW: 239.99 g/mol Boiling Pt: 176 °C Density: 2.00 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00040151 CAS Number: 2265-93-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...