2,2-Diphenylpropiononitrile ≥98.0%
Supplier: TCI America
|
Warning
|
D2534-5G
D2534-25G
TCD2534-025GEA
284
USD
TCD2534-025G
TCD2534-005G
2,2-Diphenylpropiononitrile ≥98.0%
2,2-Diphenylpropiononitrile
CAS Number: 5558-67-8
MDL Number: MFCD00001846
Molecular Formula: C15H13N
Molecular Weight: 207.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 113
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 1,1-Diphenylethyl Cyanide
Formula:
(C₆H₅)₂C(CN)CH₃ MW: 207.27 g/mol Boiling Pt: 142 °C Density: 1.07 (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001846 CAS Number: 5558-67-8 EINECS: 226-925-1 UN: 3276 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...