Di-tert-butyldichlorosilane ≥95.0%

Supplier: TCI America

D2469-5G
TCD2469-005GEA 199.62 USD
TCD2469-005G
Di-tert-butyldichlorosilane ≥95.0%
Di-tert-butyldichlorosilane

CAS Number: 18395-90-9
MDL Number: MFCD00008796
Molecular Formula: C8H18Cl2Si
Molecular Weight: 213.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Melting point (°C): -15
Flash Point (°C): 81
Specific Gravity (20/20): 1.01

Formula: C₈H₁₈Cl₂Si
MW: 213.22 g/mol
Boiling Pt: 191 °C
Melting Pt: –15 °C
Density: 1.01 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00008796
CAS Number: 18395-90-9
EINECS: 242-273-0
UN: 2987
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/93/90/28979390.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR