3,4-Dibromotoluene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,2-Dibromo-4-methylbenzene
D2462-5G
D2462-25G
TCD2462-005GEA
57
USD
TCD2462-005G
TCD2462-025G
3,4-Dibromotoluene ≥98.0%
3,4-Dibromotoluene
CAS Number: 60956-23-2
MDL Number: MFCD00079744
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 241
Melting point (°C): -10
Specific Gravity (20/20): 1.85
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 241 °C Melting Pt: –10 °C Density: 1.85 (20 °C) |
MDL Number:
MFCD00079744 CAS Number: 60956-23-2 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...