2',4'-Dichloro-5'-fluoroacetophenone
Supplier: TCI America
Synonyms:
1-(2,4-Dichloro-5-fluorophenyl)ethanone
D2385-25G
TCD2385-025GEA
36.54
USD
TCD2385-025G
2',4'-Dichloro-5'-fluoroacetophenone
2',4'-Dichloro-5'-fluoroacetophenone
CAS Number: 704-10-9
MDL Number: MFCD00077499
Molecular Formula: C8H5Cl2FO
Molecular Weight: 207.03
Form: Crystal
Color: White
Boiling point (°C): 115
Melting point (°C): 35
Flash Point (°C): 113
Formula:
C₈H₅Cl₂FO MW: 207.03 g/mol Boiling Pt: 115 °C Melting Pt: 35 °C Flash Pt: 113 °C |
MDL Number:
MFCD00077499 CAS Number: 704-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...