Diphenyliodonium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D2372-5G
TCD2372-005GEA 338 USD
TCD2372-005G
Diphenyliodonium bromide ≥98.0% (by HPLC, titration analysis)
Diphenyliodonium bromide

CAS Number: 1483-73-4
MDL Number: MFCD00031703
Molecular Formula: C12H10BrI
Molecular Weight: 361.02
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 188

Formula: C₁₂H₁₀BrI
Melting Pt: 188 °C
MDL Number: MFCD00031703
CAS Number: 1483-73-4
EINECS: 216-050-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR