Didecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2363-5G D2363-25G
TCD2363-005GEA 42 USD
TCD2363-005G TCD2363-25G
Didecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)
Didecyldimethylammonium bromide

CAS Number: 2390-68-3
MDL Number: MFCD00012194
Molecular Formula: C22H48BrN
Molecular Weight: 406.54
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Formula: C₂₂H₄₈BrN
MDL Number: MFCD00012194
CAS Number: 2390-68-3
EINECS: 219-234-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR