N,N-Dibutylformamide ≥98.0%
Supplier: TCI America
Synonyms:
Formdibutylamide
, Formic acid dibutylamide
D2147-100ML
D2147-500ML
D2147-25ML
TCD2147-500MLEA
166.46
USD
TCD2147-500ML
TCD2147-025ML
TCD2147-100ML
N,N-Dibutylformamide ≥98.0%
N,N-Dibutylformamide
CAS Number: 761-65-9
MDL Number: MFCD00003289
Molecular Formula: C9H19NO
Molecular Weight: 157.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 243
Flash Point (°C): 100
Specific Gravity (20/20): 0.88
Formula:
C₉H₁₉NO MW: 157.26 g/mol Boiling Pt: 243 °C Melting Pt: –30 °C Density: 0.88 (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003289 CAS Number: 761-65-9 EINECS: 212-090-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...