4,6-Diaminopyrimidine-2-thiol ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4,6-Diamino-2-pyrimidinethiol4,6-Diamino-2-mercaptopyrimidine

D2063-25G
TCD2063-025GEA 327.75 USD
TCD2063-025G
4,6-Diaminopyrimidine-2-thiol ≥95.0% (by titrimetric analysis)
4,6-Diaminopyrimidine-2-thiol

CAS Number: 1004-39-3
MDL Number: MFCD00006078
Molecular Formula: C4H6N4S
Molecular Weight: 142.18
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Formula: C₄H₆N₄S
MW: 142.18 g/mol
MDL Number: MFCD00006078
CAS Number: 1004-39-3
EINECS: 213-721-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR