2,5-Dimethyl-2-hexanol
Supplier: TCI America
D1968-5ML
TCD1968-005MLEA
40.6
USD
TCD1968-005ML
2,5-Dimethyl-2-hexanol
2,5-Dimethyl-2-hexanol
CAS Number: 3730-60-7
MDL Number: MFCD00021814
Molecular Formula: C8H18O
Molecular Weight: 130.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 74
Flash Point (°C): 52
Formula:
C₈H₁₈O MW: 130.23 g/mol Boiling Pt: 74 °C Density: 0.8203 g/cm³ (0 °C) Flash Pt: 52 °C |
MDL Number:
MFCD00021814 CAS Number: 3730-60-7 EINECS: 223-085-8 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...