Ethyl-3,3-diethoxypropionate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 3,3-diethoxypropanoateEthyl 3,3-diethoxypropionate

D1962-25ML D1962-100ML
TCD1962-025MLEA 108 USD
TCD1962-025ML TCD1962-100ML
Ethyl-3,3-diethoxypropionate ≥95.0%
Ethyl 3,3-diethoxypropionate

CAS Number: 10601-80-6
MDL Number: MFCD00009865
Molecular Formula: C9H18O4
Molecular Weight: 190.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 84
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 3,3-Diethoxypropionic Acid Ethyl Ester

Formula: C₉H₁₈O₄
MW: 190.24 g/mol
Boiling Pt: 86 °C
Density: 0.97 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00009865
CAS Number: 10601-80-6
EINECS: 234-223-1
https://us.vwr.com/stibo/web/std.lang.all/87/60/28978760.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR