Bis(4-fluorophenyl)methane ≥99.0%

Supplier: TCI America

Synonyms: 4,4'-Difluorodiphenylmethane

D1925-25G
TCD1925-025GEA 46 USD
TCD1925-025G
Bis(4-fluorophenyl)methane ≥99.0%
Bis(4-fluorophenyl)methane

CAS Number: 457-68-1
MDL Number: MFCD00000360
Molecular Formula: C13H10F2
Molecular Weight: 204.22
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Flash Point (°C): 134
Freezing point (°C): 28


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-fluorophenyl)methane

Formula: C₁₃H₁₀F₂
MW: 204.22
Flash Pt: 134 °C
MDL Number: MFCD00000360
CAS Number: 457-68-1
EINECS: 207-274-2
https://us.vwr.com/stibo/web/std.lang.all/87/23/28978723.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR