1-Iodo-2,4-dinitrobenzene ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dinitroiodobenzene1,3-Dinitro-4-iodobenzene

D1921-25G D1921-5G
TCD1921-25GEA 369.77 USD
TCD1921-25G TCD1921-005G
1-Iodo-2,4-dinitrobenzene ≥98.0%
1-Iodo-2,4-dinitrobenzene

CAS Number: 709-49-9
MDL Number: MFCD00039738
Molecular Formula: C6H3IN2O4
Molecular Weight: 294.00
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 87

Formula: C₆H₃IN₂O₄
MW: 294.01 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00039738
CAS Number: 709-49-9
EINECS: 211-908-3
https://us.vwr.com/stibo/web/std.lang.all/87/20/28978720.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR