2,6-Difluorobenzamide ≥97.0% (by total nitrogen basis)
Supplier: TCI America
D1907-25G
TCD1907-025GEA
30.3
USD
TCD1907-025G
2,6-Difluorobenzamide ≥97.0% (by total nitrogen basis)
2,6-Difluorobenzamide
CAS Number: 18063-03-1
MDL Number: MFCD00007972
Molecular Formula: C7H5F2NO
Molecular Weight: 157.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 145
Formula:
C₇H₅F₂NO Boiling Pt: 284 °C (1013 hPa) Melting Pt: 145 °C |
MDL Number:
MFCD00007972 CAS Number: 18063-03-1 EINECS: 241-972-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...