3,3-Diphenylpropylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3,3-Diphenylpropan-1-amine
D1855-25G
TCD1855-025GEA
87.18
USD
TCD1855-025G
3,3-Diphenylpropylamine ≥98.0% (by titrimetric analysis)
3,3-Diphenylpropylamine
CAS Number: 5586-73-2
MDL Number: MFCD00008202
Molecular Formula: C15H17N
Molecular Weight: 211.31
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 166
Flash Point (°C): 113
Freezing point (°C): 24
Other Product Names / Synonyms: alpha-(2-Aminoethyl)diphenylmethane
Formula:
C₁₅H₁₇N MW: 211.31 g/mol Boiling Pt: 166 °C Flash Pt: 113 °C |
MDL Number:
MFCD00008202 CAS Number: 5586-73-2 EINECS: 226-984-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...