(R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Supplier: TCI America
D1852-1G
TCD1852-001GEA
112
USD
TCD1852-001G
(R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
(R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
CAS Number: 69632-32-2
MDL Number: MFCD00010866
Molecular Formula: C15H13N3O5
Molecular Weight: 315.29
Form: Crystal
Color: White
Melting point (°C): 159
Formula:
C₁₅H₁₃N₃O₅ Melting Pt: 159 °C |
MDL Number:
MFCD00010866 CAS Number: 69632-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...