Dibenzosuberone ≥98.0%
Supplier: TCI America
Synonyms:
5-Dibenzosuberone
, 1-Dibenzo[b,f]cycloheptanone
, Dibenzo[b,f]cycloheptan-1-one
, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one
D1801-100G
D1801-500G
TCD1801-100GEA
84.21
USD
TCD1801-100G
TCD1801-500G
Dibenzosuberone ≥98.0%
Dibenzosuberone
CAS Number: 1210-35-1
MDL Number: MFCD00003587
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 148
Flash Point (°C): 110
Freezing point (°C): 32
Formula:
C₁₅H₁₂O MW: 208.26 g/mol Boiling Pt: 148 °C Melting Pt: 31…34 °C Density: 1.1635 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003587 CAS Number: 1210-35-1 EINECS: 214-912-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...