(3,4-Dichlorophenyl)acetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dichlorobenzyl cyanide3,4-Dichlorophenylacetonitrile

D1794-25G
TCD1794-025GEA 56 USD
TCD1794-025G
(3,4-Dichlorophenyl)acetonitrile ≥98.0%
(3,4-Dichlorophenyl)acetonitrile

CAS Number: 3218-49-3
MDL Number: MFCD00001909
Molecular Formula: C8H5Cl2N
Molecular Weight: 186.04
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 40
Flash Point (°C): 110


Other Product Names / Synonyms: 3,4-Dichlorophenylacetonitrile

Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Boiling Pt: 171 °C
Melting Pt: 40 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001909
CAS Number: 3218-49-3
EINECS: 221-743-9
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/85/79/28978579.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR