3,4-Dichlorobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (3,4-Dichlorophenyl)methanol

D1748-25G
TCD1748-025GEA 42 USD
TCD1748-025G
3,4-Dichlorobenzyl alcohol ≥98.0%
3,4-Dichlorobenzyl alcohol

CAS Number: 1805-32-9
MDL Number: MFCD00004633
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 38
Flash Point (°C): 113

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 130 °C
Melting Pt: 38 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004633
CAS Number: 1805-32-9
EINECS: 217-299-0
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/85/29/28978529.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR