3,4-Dimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,4-Dimethoxycinnamic Acid
D1728-25G
TCD1728-025GEA
45.68
USD
TCD1728-025G
3,4-Dimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
3,4-Dimethoxycinnamic acid
CAS Number: 2316-26-9
MDL Number: MFCD00004387
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 180
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(3,4-Dimethoxyphenyl)-2-propenoic Acid
Formula:
C₁₁H₁₂O₄ MW: 208.21 g/mol Boiling Pt: 343 °C (1013 hPa) Melting Pt: 180 °C |
MDL Number:
MFCD00004387 CAS Number: 2316-26-9 EINECS: 219-025-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...