1,4-Difluoro-2-nitrobenzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,5-Difluoronitrobenzene

D1700-25G
TCD1700-025GEA 142.47 USD
TCD1700-025G
1,4-Difluoro-2-nitrobenzene ≥98.0%
1,4-Difluoro-2-nitrobenzene

CAS Number: 364-74-9
MDL Number: MFCD00007054
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 90
Specific Gravity (20/20): 1.45

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 104 °C
Melting Pt: –11.7 °C
Density: 1.45 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00007054
CAS Number: 364-74-9
EINECS: 206-663-4
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/84/87/28978487.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR