2,3-Dichlorobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

D1666-25G D1666-500G
TCD1666-500GEA 284 USD
TCD1666-500G TCD1666-25G
2,3-Dichlorobenzaldehyde ≥98.0%
2,3-Dichlorobenzaldehyde

CAS Number: 6334-18-5
MDL Number: MFCD00010127
Molecular Formula: C7H4Cl2O
Molecular Weight: 175.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 62
Flash Point (°C): 135

Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 105 °C
Melting Pt: 62 °C
Density: 1.33 g/cm³ (100 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00010127
CAS Number: 6334-18-5
EINECS: 228-711-3
https://us.vwr.com/stibo/web/std.lang.all/84/61/28978461.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR