2,6-Difluorobenzonitrile ≥99.0%

Supplier: TCI America
Warning

D1650-100G D1650-25G
TCD1650-100GEA 62.84 USD
TCD1650-100G TCD1650-025G
2,6-Difluorobenzonitrile ≥99.0%
2,6-Difluorobenzonitrile

CAS Number: 1897-52-5
MDL Number: MFCD00001778
Molecular Formula: C7H3F2N
Molecular Weight: 139.10
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Flash Point (°C): 80
Freezing point (°C): 28

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 98…100 °C (31 mmHg)
Melting Pt: 30…32 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00001778
CAS Number: 1897-52-5
EINECS: 217-589-7
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/84/45/28978445.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR