2,6-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D1629-25G
TCD1629-025GEA 92.67 USD
TCD1629-025G
2,6-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
2,6-Difluorobenzoic acid

CAS Number: 385-00-2
MDL Number: MFCD00002411
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 77
Melting point (°C): 158

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 77 °C
Melting Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002411
CAS Number: 385-00-2
EINECS: 206-856-3
https://us.vwr.com/stibo/web/std.lang.all/84/23/28978423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR