2,5-Dibromothiophene ≥95.0%

Supplier: TCI America
Warning

D1549-25G D1549-100G
TCD1549-025GEA 37.54 USD
TCD1549-025G TCD1549-100G
2,5-Dibromothiophene ≥95.0%
2,5-Dibromothiophene

CAS Number: 3141-27-3
MDL Number: MFCD00005420
Molecular Formula: C4H2Br2S
Molecular Weight: 241.93
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 99
Specific Gravity (20/20): 2.14

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 210 °C
Melting Pt: –6 °C
Density: 2.14 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00005420
CAS Number: 3141-27-3
EINECS: 221-547-3
UN: 2810
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/83/57/28978357.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR