N,N-Dimethylethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: DMEA

D1538-25ML D1538-500ML
TCD1538-25MLEA 29 USD
TCD1538-25ML TCD1538-500ML
N,N-Dimethylethylamine ≥98.0% (by GC, titration analysis)
N,N-Dimethylethylamine

CAS Number: 598-56-1
MDL Number: MFCD00009039
Molecular Formula: C4H11N
Molecular Weight: 73.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 37
Flash Point (°C): -36
Specific Gravity (20/20): 0.68


Other Product Names / Synonyms: N-Ethyldimethylamine

Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 37 °C
Melting Pt: –140 °C
Density: 0.68 (20 °C)
Flash Pt: –36 °C
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8
https://us.vwr.com/stibo/web/std.lang.all/83/50/28978350.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR