Triethyl-3-phosphonopropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-(diethoxyphosphoryl)propanoate

D1524-25G D1524-250G
TCD1524-025GEA 55.83 USD
TCD1524-025G TCD1524-250G
Triethyl-3-phosphonopropionate ≥98.0%
Triethyl 3-phosphonopropionate

CAS Number: 3699-67-0
MDL Number: MFCD00015167
Molecular Formula: C9H19O5P
Molecular Weight: 238.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Ethyl 3-Diethylphosphonopropionate, 3-Phosphonopropionic Acid Triethyl Ester, 3-Diethylphosphonopropionic Acid Ethyl Ester

Formula: C₉H₁₉O₅P
MW: 238.22 g/mol
Boiling Pt: 95 °C
Density: 1.10 (20 °C)
MDL Number: MFCD00015167
CAS Number: 3699-67-0
https://us.vwr.com/stibo/web/std.lang.all/83/35/28978335.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR