4-Nitroveratrole ≥99.0%

Supplier: TCI America
Warning

Synonyms: 3,4-Dimethoxynitrobenzene

D1488-25G
TCD1488-025GEA 37.96 USD
TCD1488-025G
4-Nitroveratrole ≥99.0%
4-Nitroveratrole

CAS Number: 709-09-1
MDL Number: MFCD00007238
Molecular Formula: C8H9NO4
Molecular Weight: 183.16
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 97


Other Product Names / Synonyms: 4-Nitroveratrole

Formula: C₈H₉NO₄
MW: 183.16 g/mol
Boiling Pt: 230 °C
Melting Pt: 97 °C
Density: 1.1888 g/cm³ (132.7 °C)
MDL Number: MFCD00007238
CAS Number: 709-09-1
EINECS: 211-906-2
https://us.vwr.com/stibo/web/std.lang.all/83/11/28978311.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR