1,3-Dichloro-4-iodobenzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,4-Dichloroiodobenzene2,4-Dichloro-1-iodobenzene

D1441-25G
TCD1441-025GEA 149.21 USD
TCD1441-025G
1,3-Dichloro-4-iodobenzene ≥98.0%
1,3-Dichloro-4-iodobenzene

CAS Number: 29898-32-6
MDL Number: MFCD00001035
Molecular Formula: C6H3Cl2I
Molecular Weight: 272.89
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113


Other Product Names / Synonyms: 2,4-Dichloroiodobenzene

Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 262 °C (1013 hPa)
Density: 2.01 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001035
CAS Number: 29898-32-6
EINECS: 249-938-4
https://us.vwr.com/stibo/web/std.lang.all/82/75/28978275.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR