2,4-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D1438-10G
TCD1438-010GEA 22.78 USD
TCD1438-010G
2,4-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
2,4-Dimethylbenzoic acid

CAS Number: 611-01-8
MDL Number: MFCD00002480
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 127
Flash Point (°C): 154


Other Product Names / Synonyms: m-Xylene-4-carboxylic Acid

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 127 °C
Flash Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002480
CAS Number: 611-01-8
EINECS: 210-246-2
https://us.vwr.com/stibo/web/std.lang.all/82/72/28978272.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR