1,2-Dichloro-4-iodobenzene ≥98.0%
Supplier: TCI America
Synonyms:
3,4-Dichloroiodobenzene
D1436-10G
TCD1436-010GEA
31.47
USD
TCD1436-010G
1,2-Dichloro-4-iodobenzene ≥98.0%
1,2-Dichloro-4-iodobenzene
CAS Number: 20555-91-3
MDL Number: MFCD00019014
Molecular Formula: C6H3Cl2I
Molecular Weight: 272.89
Purity/Analysis Method:
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30
Other Product Names / Synonyms: 3,4-Dichloroiodobenzene
Formula:
C₆H₃Cl₂I MW: 272.9 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00019014 CAS Number: 20555-91-3 EINECS: 243-874-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...