2,3-Dihydrofuran ≥98.0%
Supplier: TCI America
D1415-500ML
D1415-25ML
D1415-100ML
TCD1415-25MLEA
46.1
USD
TCD1415-25ML
TCD1415-500ML
TCD1415-100ML
2,3-Dihydrofuran ≥98.0%
2,3-Dihydrofuran
CAS Number: 1191-99-7
MDL Number: MFCD00003205
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 54
Flash Point (°C): -25
Specific Gravity (20/20): 0.93
Formula:
C₄H₆O Boiling Pt: 54 °C Density: 0.93 (20 °C) Flash Pt: –25 °C |
MDL Number:
MFCD00003205 CAS Number: 1191-99-7 EINECS: 214-747-7 UN: 3271 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...