(±)-δ-Dodecalactone ≥98.0%
Supplier: TCI America
Synonyms:
5-Dodecanolide
, δ-Dodecalactone
, 6-Heptyltetrahydro-2H-pyran-2-one
, δ-Dodecanolactone
, (±)-Tetrahydro-6-heptyl-2H-pyran-2-one
, (±)-Dodecan-5-olide
D1414-25ML
TCD1414-025MLEA
25.38
USD
TCD1414-025ML
(±)-δ-Dodecalactone ≥98.0%
(±)-δ-Dodecalactone
CAS Number: 713-95-1
MDL Number: MFCD00006651
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 165
Specific Gravity (20/20): 0.95
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: delta-Dodecalactone, delta-Laurolactone
Formula:
C₁₂H₂₂O₂ MW: 198.31 g/mol Boiling Pt: 130 °C Melting Pt: –7 °C Density: 0.95 (20 °C) Flash Pt: 165 °C |
MDL Number:
MFCD00006651 CAS Number: 713-95-1 EINECS: 211-932-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...