Chloro(methyl)(diphenyl)silane ≥95.0%

Supplier: TCI America

D1390-25ML
TCD1390-025MLEA 89.23 USD
TCD1390-025ML
Chloro(methyl)(diphenyl)silane ≥95.0%
Chloro(methyl)(diphenyl)silane

CAS Number: 144-79-6
MDL Number: MFCD00000498
Molecular Formula: C13H13ClSi
Molecular Weight: 232.78
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 295
Melting point (°C): -22
Flash Point (°C): 141
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Chlorodiphenylmethylsilane, Chloromethyldiphenylsilane

Formula: C₁₃H₁₃ClSi
Boiling Pt: 295 °C
Melting Pt: –22 °C
Density: 1.11 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00000498
CAS Number: 144-79-6
EINECS: 205-639-0
UN: 2987
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/82/38/28978238.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR