N,N-Diethylhydroxylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1339-500ML
D1339-25ML
D1339-100ML
TCD1339-500MLEA
102.52
USD
TCD1339-500ML
TCD1339-25ML
TCD1339-100ML
N,N-Diethylhydroxylamine ≥98.0% (by GC, titration analysis)
N,N-Diethylhydroxylamine
CAS Number: 3710-84-7
MDL Number: MFCD00002126
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 51
Specific Gravity (20/20): 0.87
Formula:
C₄H₁₁NO Boiling Pt: 122 °C Density: 0.87 (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00002126 CAS Number: 3710-84-7 EINECS: 223-055-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...