Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥98.0% (by total nitrogen basis)
Supplier: TCI America
D1328-25G
D1328-500G
TCD1328-500GEA
255
USD
TCD1328-500G
TCD1328-025G
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥98.0% (by total nitrogen basis)
Diuron (N-(3,4-Dichlorophenyl)-N',N'-dimethylurea)
CAS Number: 330-54-1
MDL Number: MFCD00018136
Molecular Formula: C9H10Cl2N2O
Molecular Weight: 233.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 159
Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: Diuron
Formula:
C₉H₁₀Cl₂N₂O Melting Pt: 159 °C |
MDL Number:
MFCD00018136 CAS Number: 330-54-1 EINECS: 206-354-4 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...