1,9-Decadiyne ≥98.0%

Supplier: TCI America

D1326-5ML
TCD1326-005MLEA 136.92 USD
TCD1326-005ML
1,9-Decadiyne ≥98.0%
1,9-Decadiyne

CAS Number: 1720-38-3
MDL Number: MFCD00014926
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 65
Specific Gravity (20/20): 0.82

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 72 °C
Melting Pt: –10.15 °C
Density: 0.82 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00014926
CAS Number: 1720-38-3
EINECS: 217-012-9
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR