Dimethyl selenide ≥98.0%
|
Danger
|
D1289-1G
TCD1289-001GEA
143.17
USD
TCD1289-001G
Dimethyl selenide ≥98.0%
Dimethyl selenide
CAS Number: 593-79-3
MDL Number: MFCD00014848
Molecular Formula: C2H6Se
Molecular Weight: 109.03
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 58
Specific Gravity (20/20): 1.43
Product Categories: Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Dimethylselenium
Formula:
C₂H₆Se MW: 109.03 g/mol Boiling Pt: 58 °C Density: 1.43 (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014848 CAS Number: 593-79-3 EINECS: 209-807-4 UN: 3282 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...