1,3-Dibromo-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione

D1265-25G D1265-500G
TCD1265-500GEA 180.67 USD
TCD1265-500G TCD1265-25G
1,3-Dibromo-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)
1,3-Dibromo-5,5-dimethylhydantoin

CAS Number: 77-48-5
MDL Number: MFCD00003189
Molecular Formula: C5H6Br2N2O2
Molecular Weight: 285.92
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Boiling point (°C): 376
Melting point (°C): 193


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Dibromantin, DBDMH

Formula: C₅H₆Br₂N₂O₂
MW: 285.92 g/mol
Boiling Pt: 376 °C
Melting Pt: 193 °C
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9
https://us.vwr.com/stibo/web/std.lang.all/81/58/28978158.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR