6-Methylquinaldine ≥99.0%
Supplier: TCI America
Synonyms:
2,6-Dimethylquinoline
D1236-25G
D1236-5G
TCD1236-025GEA
324.8
USD
TCD1236-025G
TCD1236-5G
6-Methylquinaldine ≥99.0%
6-Methylquinaldine
CAS Number: 877-43-0
MDL Number: MFCD00006762
Molecular Formula: C11H11N
Molecular Weight: 157.22
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 58
Formula:
C₁₁H₁₁N MW: 157.22 g/mol Boiling Pt: 267 °C Melting Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006762 CAS Number: 877-43-0 EINECS: 212-891-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...