6-Dodecyne ≥98.0%

Supplier: TCI America

D1192-5ML
TCD1192-005MLEA 68.65 USD
TCD1192-005ML
6-Dodecyne ≥98.0%
6-Dodecyne

CAS Number: 6975-99-1
MDL Number: MFCD00041658
Molecular Formula: C12H22
Molecular Weight: 166.31
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 209
Flash Point (°C): 65
Specific Gravity (20/20): 0.78

Formula: C₁₂H₂₂
Boiling Pt: 209 °C
Density: 0.78 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00041658
CAS Number: 6975-99-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR